Assay Details
Target Gene Details
Entrez Gene ID: | 51001 |
Gene Name: | mitochondrial transcription termination factor 3 |
Gene Aliases: |
CGI-12, MTERFD1 |
Location: |
Chr.8:96239398-96261613 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTERF3 | NM_001286643.1 | 2 | 204 | NP_001273572.1 |
| NM_015942.4 | 2 | 204 | NP_057026.3 | |
| XM_006716570.3 | 2 | 197 | XP_006716633.1 | |
| AK309002.1 | 2 | 201 | ||
| BC012995.2 | 2 | 159 | AAH12995.2 | |
| HY116693.1 | 2 | 204 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020494 | Chr.8:96188447 - 96286981 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map