Assay Details
Target Gene Details
Entrez Gene ID: | 80162 |
Gene Name: | protein-glucosylgalactosylhydroxylysine glucosidase |
Gene Aliases: |
ATHL1 |
Location: |
Chr.11:287760-295688 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGGHG | NM_025092.4 | 14 | 2715 | NP_079368.3 |
| XM_011520383.2 | 17 | 3641 | XP_011518685.1 | |
| XM_011520384.2 | 16 | 3698 | XP_011518686.1 | |
| XM_017018355.1 | 15 | 3093 | XP_016873844.1 | |
| AK026288.1 | 8 | 1357 | BAB15431.1 | |
| AK074155.1 | 11 | 3877 | BAB84981.1 | |
| AK090428.1 | 8 | 2332 | BAC03409.1 | |
| AK296137.1 | 10 | 1594 | ||
| AL832932.1 | 7 | 1373 | CAH56316.1 | |
| BC109257.2 | 11 | 2114 | AAI09258.1 | |
| BC109258.2 | 11 | 2114 | AAI09259.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891905 | Chr.11:269147 - 336332 on Build GRCh38 | Gain |
|
| nsv516809 | Chr.11:290233 - 295876 on Build GRCh38 | Loss |
|
| nsv552765 | Chr.11:199256 - 320836 on Build GRCh38 | Gain |
|
| dgv1505n54 | Chr.11:198510 - 470331 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map