Assay Details
Target Gene Details
Entrez Gene ID: | 90196 |
Gene Name: | Sys1 golgi trafficking protein |
Gene Aliases: |
C20orf169, dJ453C12.4, dJ453C12.4.1 |
Location: |
Chr.20:45361937-45376802 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYS1 | NM_001099791.2 | NP_001093261.1 | ||
| NM_001197129.1 | 5 | 2372 | NP_001184058.1 | |
| NM_033542.3 | 4 | 2348 | NP_291020.1 | |
| AI041176.1 | ||||
| AK075264.1 | 4 | 2311 | BAC11506.1 | |
| AK172738.1 | ||||
| AL591714.1 | 4 | 2243 | CAC39448.1 |
Target Gene Details
Entrez Gene ID: | 767557 |
Gene Name: | SYS1-DBNDD2 readthrough (NMD candidate) |
Gene Aliases: |
C20orf169-DBNDD2 |
Location: |
Chr.20:45363169-45410610 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYS1-DBNDD2 | NR_003189.2 | |||
| BI199175.1 | ||||
| DN994280.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map