Assay Details
Target Gene Details
Entrez Gene ID: | 57521 |
Gene Name: | regulatory associated protein of MTOR complex 1 |
Gene Aliases: |
KOG1, Mip1 |
Location: |
Chr.17:80544825-80966373 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPTOR | NM_001163034.1 | 1 | 649 | NP_001156506.1 |
| NM_020761.2 | 1 | 649 | NP_065812.1 | |
| AK055912.1 | 1 | 495 | ||
| AK295964.1 | 1 | 655 | ||
| AK296321.1 | 1 | 643 | ||
| AY090663.1 | 1 | 206 | AAM09075.1 | |
| BC064515.1 | 1 | 597 | AAH64515.1 | |
| BC136652.1 | 1 | 39 | ||
| BC136654.1 | 1 | 39 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056989 | Chr.17:80534340 - 80557095 on Build GRCh38 | Gain |
|
| esv24254 | Chr.17:80396528 - 80656857 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs144378496] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map