Assay Details
Target Gene Details
Entrez Gene ID: | 11163 |
Gene Name: | nudix hydrolase 4 |
Gene Aliases: |
DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P |
Location: |
Chr.12:93377923-93403248 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUDT4 | NM_001301022.1 | NP_001287951.1 | ||
| NM_001301023.1 | NP_001287952.1 | |||
| NM_001301024.1 | NP_001287953.1 | |||
| NM_019094.5 | NP_061967.3 | |||
| NM_199040.3 | NP_950241.1 | |||
| XM_011537794.2 | XP_011536096.1 | |||
| XM_011537795.2 | XP_011536097.1 | |||
| XM_011537796.2 | XP_011536098.1 | |||
| AB007956.2 | ||||
| AF067803.1 | AAF75563.1 | |||
| AF191649.1 | AAF68855.1 | |||
| AF191650.1 | AAF68856.1 | |||
| AF191651.1 | AAF68857.1 | |||
| AF191652.2 | AAF68858.2 | |||
| AK223497.1 | BAD97217.1 | |||
| AK304296.1 | ||||
| AK315778.1 | ||||
| BC012069.1 | AAH12069.1 | |||
| BC051310.1 | AAH51310.1 | |||
| BP315657.1 | ||||
| CN335956.1 | ||||
| CR749445.1 | CAH18283.1 | |||
| DC323957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044304 | Chr.12:93219056 - 93419239 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map