Assay Details
Target Gene Details
Entrez Gene ID: | 4927 |
Gene Name: | nucleoporin 88 |
Gene Aliases: |
- |
Location: |
Chr.17:5384833-5419739 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP88 | NM_001320653.1 | 1 | 199 | NP_001307582.1 |
| NM_002532.5 | 1 | 199 | NP_002523.2 | |
| XM_011523893.2 | 1 | 122 | XP_011522195.1 | |
| XM_017024694.1 | 1 | 122 | XP_016880183.1 | |
| AK225247.1 | 1 | 122 | ||
| AK298158.1 | ||||
| AK299000.1 | 1 | 122 | ||
| AK308142.1 | ||||
| BC000335.2 | 1 | 112 | AAH00335.1 | |
| BI552947.1 | 1 | 141 | ||
| BX956129.1 | 1 | 199 | ||
| Y08612.2 | 1 | 158 | CAA69904.1 |
Target Gene Details
Entrez Gene ID: | 84268 |
Gene Name: | RPA interacting protein |
Gene Aliases: |
HRIP, RIP |
Location: |
Chr.17:5419185-5433020 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPAIN |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map