Assay Details
Target Gene Details
Entrez Gene ID: | 1316 |
Gene Name: | Kruppel like factor 6 |
Gene Aliases: |
BCD1, CBA1, COPEB, CPBP, GBF, PAC1, ST12, ZF9 |
Location: |
Chr.10:3775996-3785281 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF6 | NM_001160124.1 | 1 | 130 | NP_001153596.1 |
| NM_001160125.1 | 1 | 130 | NP_001153597.1 | |
| NM_001300.5 | 1 | 130 | NP_001291.3 | |
| NR_027653.1 | 1 | 130 | ||
| AB017493.1 | BAA33050.1 | |||
| AK293259.1 | 1 | 64 | ||
| AK316596.1 | 1 | 58 | ||
| BC000311.2 | AAH00311.1 | |||
| BM544849.1 | 1 | 131 | ||
| BP357332.1 | 1 | 130 | ||
| CR975091.1 | ||||
| CR994352.1 | ||||
| U51869.1 | AAB94792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv661n100 | Chr.10:3749938 - 3788015 on Build GRCh38 | Gain |
|
| nsv1126240 | Chr.10:3785108 - 3785708 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map