Assay Details
Target Gene Details
Entrez Gene ID: | 28968 |
Gene Name: | solute carrier family 6 member 16 |
Gene Aliases: |
NT5, NTT5 |
Location: |
Chr.19:49289632-49332320 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A16 | NM_014037.2 | 12 | 2470 | NP_054756.2 |
| XM_005258820.2 | 13 | 2571 | XP_005258877.1 | |
| XM_006723168.3 | 12 | 2486 | XP_006723231.1 | |
| XM_011526859.2 | 11 | 2393 | XP_011525161.2 | |
| XM_011526860.2 | 13 | 2783 | XP_011525162.1 | |
| XM_011526861.2 | 11 | 1893 | XP_011525163.1 | |
| XM_017026712.1 | 12 | 2486 | XP_016882201.1 | |
| AF151977.1 | 11 | 2278 | AAD38044.1 | |
| AF265578.1 | AAG41362.1 | |||
| AL136856.1 | 11 | 2282 | CAB66790.1 | |
| BC034948.1 | 11 | 2650 | AAH34948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| esv2658176 | Chr.19:49287651 - 49297841 on Build GRCh38 | Deletion |
|
| dgv52n68 | Chr.19:49286685 - 49456131 on Build GRCh38 | Loss |
|
| esv3644620 | Chr.19:49287578 - 49297780 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map