Assay Details
Target Gene Details
Entrez Gene ID: | 645432 |
Gene Name: | arrestin domain containing 5 |
Gene Aliases: |
- |
Location: |
Chr.19:4867614-4915469 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARRDC5 | XM_011528181.2 | XP_011526483.1 |
Target Gene Details
Entrez Gene ID: | 29128 |
Gene Name: | ubiquitin like with PHD and ring finger domains 1 |
Gene Aliases: |
ICBP90, Np95, RNF106, TDRD22, hNP95, hUHRF1, huNp95 |
Location: |
Chr.19:4903080-4962154 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UHRF1 | NM_001290050.1 | NP_001276979.1 | ||
| AB177623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065556 | Chr.19:4844193 - 4945238 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73919353] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map