Assay Details
Target Gene Details
Entrez Gene ID: | 2042 |
Gene Name: | EPH receptor A3 |
Gene Aliases: |
EK4, ETK, ETK1, HEK, HEK4, TYRO4 |
Location: |
Chr.3:89107524-89482134 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA3 | NM_005233.5 | 1 | 243 | NP_005224.2 |
| NM_182644.2 | 1 | 243 | NP_872585.1 | |
| XM_005264715.2 | 1 | 168 | XP_005264772.1 | |
| XM_005264716.2 | 1 | 238 | XP_005264773.1 | |
| AF213459.1 | 1 | 243 | AAG43576.1 | |
| AF213460.1 | 1 | 105 | AAG43577.1 | |
| AK291411.1 | 1 | 76 | ||
| BC063282.1 | 1 | 147 | AAH63282.1 | |
| M83941.1 | 1 | 118 | AAA58633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834760 | Chr.3:89036379 - 89250248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map