Assay Details
Target Gene Details
Entrez Gene ID: | 105378553 |
Gene Name: | lncRNA antisense to PTPRE protein-coding gene |
Gene Aliases: |
- |
Location: |
Chr.10:128057732-128062425 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AS-PTPRE | NR_136149.1 | |||
| NR_136150.1 | ||||
| KR186191.1 | ||||
| KR186192.1 | ||||
| KR186193.1 | ||||
| KR186194.1 | ||||
| KR186195.1 |
Target Gene Details
Entrez Gene ID: | 5791 |
Gene Name: | protein tyrosine phosphatase, receptor type E |
Gene Aliases: |
HPTPE, PTPE, R-PTP-EPSILON |
Location: |
Chr.10:127907061-128085855 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv989n100 | Chr.10:128040003 - 128445808 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map