Assay Details
Target Gene Details
Entrez Gene ID: | 1915 |
Gene Name: | eukaryotic translation elongation factor 1 alpha 1 |
Gene Aliases: |
CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1 |
Location: |
Chr.6:73515750-73521032 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EEF1A1 | NM_001402.5 | 8 | 2411 | NP_001393.1 |
| AJ420488.1 | 7 | 2371 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2763576 | Chr.6:73487915 - 73567526 on Build GRCh38 | Gain |
|
| nsv5350 | Chr.6:73512272 - 73557365 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map