Assay Details
Target Gene Details
Entrez Gene ID: | 55268 |
Gene Name: | enoyl-CoA hydratase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.1:52895910-52927212 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001535 | Chr.1:52620368 - 53346539 on Build GRCh38 | Gain |
|
| nsv829859 | Chr.1:52723470 - 52925273 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map