Assay Details
Target Gene Details
Entrez Gene ID: | 81555 |
Gene Name: | Yip1 domain family member 5 |
Gene Aliases: |
FinGER5, SB140, SMAP-5, SMAP5, YIP1A |
Location: |
Chr.5:144158159-144170714 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF5 | NM_001024947.3 | 2 | 504 | NP_001020118.1 |
| NM_001271732.1 | NP_001258661.1 | |||
| NM_030799.8 | 2 | 253 | NP_110426.4 | |
| AF140225.1 | 1 | 63 | AAG48521.1 | |
| AF318329.1 | 2 | 142 | AAL55836.1 | |
| AK054576.1 | 2 | 190 | BAB70763.1 | |
| AK124065.1 | 2 | 411 | ||
| AK223515.1 | 2 | 116 | BAD97235.1 | |
| AK307251.1 | 2 | 116 | ||
| AY037152.1 | 2 | 185 | AAK67644.1 | |
| AY358863.1 | 2 | 137 | AAQ89222.1 | |
| AY640925.1 | 2 | 169 | AAV51256.1 | |
| AY640926.1 | 2 | 169 | AAV51257.1 | |
| AY640927.1 | 2 | 169 | AAV51258.1 | |
| AY640928.1 | 2 | 420 | AAV51259.1 | |
| AY640929.1 | ||||
| AY640934.1 | 2 | 169 | AAV51260.1 | |
| BC007829.2 | 2 | 171 | AAH07829.1 | |
| BC014253.2 | 2 | 191 | AAH14253.1 | |
| BC024737.1 | 2 | 188 | AAH24737.1 | |
| BP310538.1 | 2 | 253 | ||
| BX647859.1 | 2 | 192 | ||
| CR749463.1 | 2 | 192 | CAH18295.1 |
More Information
Set Membership: |
|

Genomic Map