Assay Details
Target Gene Details
Entrez Gene ID: | 1902 |
Gene Name: | lysophosphatidic acid receptor 1 |
Gene Aliases: |
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1 |
Location: |
Chr.9:110873252-111039259 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
More Information
Set Membership: |
|

Genomic Map