Assay Details
Target Gene Details
Entrez Gene ID: | 55812 |
Gene Name: | spermatogenesis associated 7 |
Gene Aliases: |
HEL-S-296, HSD-3.1, HSD3, LCA3 |
Location: |
Chr.14:88385638-88470350 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATA7 | NM_001040428.3 | 5 | 689 | NP_001035518.1 |
| NM_018418.4 | 6 | 785 | NP_060888.2 | |
| XM_005267851.1 | 6 | 756 | XP_005267908.1 | |
| XM_005267852.1 | 5 | 685 | XP_005267909.1 | |
| XM_005267854.1 | 7 | 818 | XP_005267911.1 | |
| XM_005267855.1 | 6 | 751 | XP_005267912.1 | |
| XM_006720204.1 | 6 | 756 | XP_006720267.1 | |
| XM_006720205.1 | 6 | 756 | XP_006720268.1 | |
| XM_011536951.1 | 6 | 950 | XP_011535253.1 | |
| XM_011536952.1 | 4 | 568 | XP_011535254.1 | |
| XM_011536953.1 | 5 | 889 | XP_011535255.1 | |
| XM_017021452.1 | 6 | 950 | XP_016876941.1 | |
| XM_017021453.1 | 6 | 916 | XP_016876942.1 | |
| XM_017021454.1 | 7 | 818 | XP_016876943.1 | |
| XM_017021455.1 | 6 | 751 | XP_016876944.1 | |
| XM_017021456.1 | 7 | 764 | XP_016876945.1 | |
| XM_017021457.1 | 5 | 889 | XP_016876946.1 | |
| AF144487.3 | 6 | 765 | AAF66077.2 | |
| AF144488.2 | 4 | 490 | AAF66078.1 | |
| AK055864.1 | 6 | 707 | ||
| AK225746.1 | 7 | 756 | ||
| AK290631.1 | 5 | 660 | ||
| AK293007.1 | 7 | 738 | ||
| AL136604.1 | 5 | 669 | CAB66539.1 | |
| BC008656.1 | 1 | 217 | AAH08656.2 | |
| BC090875.1 | 6 | 1072 | AAH90875.1 | |
| CR533563.1 | 5 | 514 | CAG38594.1 | |
| CR936777.1 | 6 | 731 | ||
| DA568174.1 | ||||
| EU668353.1 | 6 | 761 |
More Information
Set Membership: |
|

Genomic Map