Assay Details
Target Gene Details
Entrez Gene ID: | 2780 |
Gene Name: | G protein subunit alpha transducin 2 |
Gene Aliases: |
ACHM4, GNATC |
Location: |
Chr.1:109603091-109619733 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAT2 | NM_005272.3 | NP_005263.1 | ||
| XM_011541264.2 | XP_011539566.1 | |||
| AF493909.1 | AAM12623.1 | |||
| BC000233.1 | AAH00233.1 | |||
| BT009844.1 | AAP88846.1 |
More Information
Set Membership: |
|

Genomic Map