Assay Details
Target Gene Details
Entrez Gene ID: | 113451 |
Gene Name: | antizyme inhibitor 2 |
Gene Aliases: |
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp |
Location: |
Chr.1:33081110-33162288 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AZIN2 | NM_001293562.1 | NP_001280491.1 | ||
| NM_001301823.1 | 1 | 281 | NP_001288752.1 | |
| NM_001301824.1 | 1 | 281 | NP_001288753.1 | |
| NM_052998.3 | NP_443724.1 | |||
| XM_005270404.2 | XP_005270461.1 | |||
| XM_005270406.1 | XP_005270463.1 | |||
| XM_005270407.1 | XP_005270464.1 | |||
| XM_006710312.1 | XP_006710375.1 | |||
| XM_011540557.1 | XP_011538859.1 | |||
| XM_011540563.1 | XP_011538865.1 | |||
| XM_011540566.1 | XP_011538868.1 | |||
| XM_017000167.1 | XP_016855656.1 | |||
| XM_017000168.1 | XP_016855657.1 | |||
| XM_017000169.1 | XP_016855658.1 | |||
| XM_017000170.1 | 1 | 108 | XP_016855659.1 | |
| XM_017000171.1 | XP_016855660.1 | |||
| XM_017000172.1 | XP_016855661.1 | |||
| XM_017000173.1 | XP_016855662.1 | |||
| XM_017000174.1 | XP_016855663.1 | |||
| XM_017000175.1 | XP_016855664.1 | |||
| AK057051.1 | 1 | 255 | BAB71356.1 | |
| AK095127.1 | 1 | 262 | BAC04489.1 | |
| AK315677.1 | ||||
| AL832640.2 | ||||
| AY325129.1 | AAQ62560.1 | |||
| BC010449.1 | AAH10449.1 | |||
| BC028128.1 | AAH28128.1 | |||
| DB456925.1 | 1 | 281 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576805 | Chr.1:32896710 - 33106481 on Build GRCh38 | Gain |
|
| nsv834602 | Chr.1:32947691 - 33116221 on Build GRCh38 | Loss |
|
| nsv545962 | Chr.1:32982619 - 33129611 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map