Assay Details
Target Gene Details
Entrez Gene ID: | 51649 |
Gene Name: | mitochondrial ribosomal protein S23 |
Gene Aliases: |
CGI-138, HSPC329, MRP-S23 |
Location: |
Chr.17:57838926-57850072 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS23 | NM_016070.3 | 5 | 806 | NP_057154.2 |
| AF151896.1 | 5 | 753 | AAD34133.1 | |
| AF161447.1 | 5 | 761 | AAF29007.1 | |
| BC000242.1 | 5 | 772 | AAH00242.1 |
More Information
Set Membership: |
|

Genomic Map