Assay Details
Target Gene Details
Entrez Gene ID: | 1387 |
Gene Name: | CREB binding protein |
Gene Aliases: |
CBP, KAT3A, RSTS |
Location: |
Chr.16:3725054-3880727 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREBBP | NM_001079846.1 | 30 | 7700 | NP_001073315.1 |
| NM_004380.2 | 31 | 7814 | NP_004371.2 | |
| XM_005255124.4 | 30 | 8376 | XP_005255181.1 | |
| XM_005255125.4 | 30 | 8004 | XP_005255182.1 | |
| XM_006720848.3 | 29 | 8160 | XP_006720911.1 | |
| XM_011522381.2 | 30 | 7303 | XP_011520683.1 | |
| XM_017022944.1 | 31 | 8415 | XP_016878433.1 | |
| BC036486.1 | 1 | 528 | AAH36486.1 | |
| U47741.1 | 32 | 7808 | AAC51770.1 | |
| U89355.1 | 5 | 3081 | AAC51340.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051657 | Chr.16:3644759 - 3788297 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map