Assay Details
Target Gene Details
Entrez Gene ID: | 594857 |
Gene Name: | neuropeptide S |
Gene Aliases: |
- |
Location: |
Chr.10:127549349-127552671 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPS | NM_001030013.1 | NP_001025184.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7607 | Chr.10:127528825 - 127573934 on Build GRCh38 | Deletion |
|
| nsv1048716 | Chr.10:127524465 - 127552592 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map