Assay Details
Target Gene Details
Entrez Gene ID: | 7040 |
Gene Name: | transforming growth factor beta 1 |
Gene Aliases: |
CED, DPD1, LAP, TGFB, TGFbeta |
Location: |
Chr.19:41330531-41353933 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TGFB1 | NM_000660.5 | 1 | 315 | NP_000651.3 |
| XM_011527242.1 | 1 | 308 | XP_011525544.1 | |
| AK307742.1 | ||||
| BG115704.1 | 1 | 315 | ||
| DA935180.1 | 1 | 308 | ||
| X02812.1 | 1 | 265 | CAA26580.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521311 | Chr.19:41350981 - 41376807 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map