Assay Details
Target Gene Details
Entrez Gene ID: | 51154 |
Gene Name: | MRT4 homolog, ribosome maturation factor |
Gene Aliases: |
C1orf33, MRT4, dJ657E11.4 |
Location: |
Chr.1:19251581-19260128 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRTO4 | NM_016183.3 | 8 | 1046 | NP_057267.2 |
| XM_006710675.3 | 8 | 1204 | XP_006710738.1 | |
| AF173378.1 | 8 | 821 | AAD52608.1 | |
| AK024227.1 | 8 | 807 | ||
| AK027569.1 | 8 | 822 | BAB55205.1 | |
| AK295328.1 | 7 | 1101 | ||
| AY303790.1 | 8 | 802 | AAP68821.1 | |
| BC003013.1 | 8 | 820 | AAH03013.1 | |
| BC006504.2 | 8 | 820 | AAH06504.1 |
More Information
Set Membership: |
|

Genomic Map