Assay Details
Target Gene Details
Entrez Gene ID: | 51174 |
Gene Name: | tubulin delta 1 |
Gene Aliases: |
TUBD |
Location: |
Chr.17:59859467-59892948 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBD1 | NM_001193609.1 | 6 | 1126 | NP_001180538.1 |
| NM_001193610.1 | 7 | 1291 | NP_001180539.1 | |
| NM_001193611.1 | NP_001180540.1 | |||
| NM_001193612.1 | 7 | 1222 | NP_001180541.1 | |
| NM_001193613.1 | 4 | 715 | NP_001180542.1 | |
| NM_016261.3 | 7 | 1291 | NP_057345.2 | |
| XM_005257425.1 | 6 | 1070 | XP_005257482.1 | |
| XM_011524871.2 | XP_011523173.1 | |||
| XM_017024716.1 | 7 | 1289 | XP_016880205.1 | |
| XM_017024717.1 | XP_016880206.1 | |||
| XM_017024718.1 | XP_016880207.1 | |||
| XM_017024719.1 | 5 | 912 | XP_016880208.1 | |
| XM_017024720.1 | 4 | 701 | XP_016880209.1 | |
| AB030837.1 | 6 | 1004 | BAD16590.1 | |
| AB031044.1 | 5 | 839 | BAD83580.1 | |
| AB031045.1 | BAD83581.1 | |||
| AF201333.1 | 6 | 1003 | AAF09584.1 | |
| AK022771.1 | 8 | 1181 | ||
| AK024096.1 | 3 | 301 | BAB14825.1 | |
| AK024741.1 | 4 | 551 | ||
| AK298492.1 | 7 | 1204 | ||
| AK301312.1 | 5 | 663 | ||
| AK315172.1 | 7 | 1218 | ||
| BC000258.1 | 6 | 1101 | AAH00258.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3640914 | Chr.17:59863061 - 59886224 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map