Assay Details
Target Gene Details
Entrez Gene ID: | 1910 |
Gene Name: | endothelin receptor type B |
Gene Aliases: |
ABCDS, ET-B, ET-BR, ETB, ETB1, ETBR, ETRB, HSCR, HSCR2, WS4A |
Location: |
Chr.13:77895481-77975529 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDNRB | NM_000115.3 | 8 | 3987 | NP_000106.1 |
| NM_001122659.2 | 7 | 4011 | NP_001116131.1 | |
| NM_001201397.1 | 8 | 4176 | NP_001188326.1 | |
| NM_003991.3 | 7 | 2235 | NP_003982.1 | |
| AB209198.1 | 7 | 3990 | BAD92435.1 | |
| D90402.1 | 8 | 3990 | BAA14398.1 | |
| H28710.1 | ||||
| S57283.1 | 8 | 3990 | AAB25531.1 |
Target Gene Details
Entrez Gene ID: | 100505518 |
Gene Name: | EDNRB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:77818937-77908442 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDNRB-AS1 | NR_103853.1 |
More Information
Set Membership: |
|

Genomic Map