Assay Details
Target Gene Details
Entrez Gene ID: | 8500 |
Gene Name: | PTPRF interacting protein alpha 1 |
Gene Aliases: |
LIP.1, LIP1, LIPRIN |
Location: |
Chr.11:70270687-70384501 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPFIA1 | NM_003626.3 | 2 | 361 | NP_003617.1 |
| NM_177423.2 | 2 | 361 | NP_803172.1 | |
| NR_045286.1 | 2 | 361 | ||
| XM_006718716.3 | 2 | 374 | XP_006718779.1 | |
| XM_011545306.2 | 2 | 246 | XP_011543608.1 | |
| XM_011545307.2 | 2 | 374 | XP_011543609.1 | |
| XM_011545308.2 | 2 | 374 | XP_011543610.1 | |
| XM_011545309.2 | 2 | 374 | XP_011543611.1 | |
| XM_011545310.2 | 2 | 374 | XP_011543612.1 | |
| XM_011545311.2 | 2 | 374 | XP_011543613.1 | |
| XM_011545312.2 | 2 | 374 | XP_011543614.1 | |
| XM_011545313.2 | 2 | 374 | XP_011543615.1 | |
| XM_011545314.2 | 2 | 374 | XP_011543616.1 | |
| XM_011545315.2 | 2 | 374 | XP_011543617.1 | |
| XM_011545316.2 | 2 | 374 | XP_011543618.1 | |
| XM_011545317.2 | 2 | 374 | XP_011543619.1 | |
| XM_011545319.1 | 2 | 374 | XP_011543621.1 | |
| XM_017018448.1 | 2 | 374 | XP_016873937.1 | |
| XM_017018449.1 | 2 | 374 | XP_016873938.1 | |
| XM_017018450.1 | 2 | 374 | XP_016873939.1 | |
| AB209314.1 | 2 | 288 | BAD92551.1 | |
| AW163189.1 | 2 | 361 | ||
| BC034046.1 | 2 | 324 | AAH34046.1 | |
| U22815.1 | 2 | 375 | AAC50172.1 | |
| U22816.1 | 2 | 375 | AAC50173.1 |
More Information
Set Membership: |
|

Genomic Map