Assay Details
Target Gene Details
Entrez Gene ID: | 2004 |
Gene Name: | ELK3, ETS transcription factor |
Gene Aliases: |
ERP, NET, SAP-2, SAP2 |
Location: |
Chr.12:96194382-96269835 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELK3 | NM_005230.3 | NP_005221.2 | ||
| XM_006719275.3 | XP_006719338.1 | |||
| AK312694.1 | ||||
| BC017371.1 | AAH17371.1 | |||
| DA754301.1 | ||||
| Z36715.1 | CAA85309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559841 | Chr.12:96194030 - 96194815 on Build GRCh38 | Loss |
|
| dgv2807n54 | Chr.12:96194215 - 96195012 on Build GRCh38 | Loss |
|
| dgv2806n54 | Chr.12:96194111 - 96195063 on Build GRCh38 | Gain |
|
| esv26105 | Chr.12:96194003 - 96195094 on Build GRCh38 | Loss |
|
| nsv559846 | Chr.12:96194267 - 96194815 on Build GRCh38 | Loss |
|
| nsv559842 | Chr.12:96194083 - 96197874 on Build GRCh38 | Gain |
|
| nsv559848 | Chr.12:96194447 - 96195063 on Build GRCh38 | Gain |
|
| dgv2805n54 | Chr.12:96193146 - 96195063 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112402795] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map