Assay Details
Target Gene Details
Entrez Gene ID: | 23554 |
Gene Name: | tetraspanin 12 |
Gene Aliases: |
EVR5, NET-2, NET2, TM4SF12 |
Location: |
Chr.7:120787320-120858369 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPAN12 | NM_012338.3 | NP_036470.1 | ||
| XM_005250239.2 | XP_005250296.1 | |||
| XM_011515993.1 | XP_011514295.1 | |||
| XM_011515994.1 | XP_011514296.1 | |||
| XM_017011912.1 | XP_016867401.1 | |||
| XM_017011913.1 | XP_016867402.1 | |||
| XM_017011914.1 | XP_016867403.1 | |||
| AF124522.1 | AAD17317.1 | |||
| AK299247.1 | ||||
| AK307161.1 | ||||
| AK312239.1 | ||||
| AY358703.1 | AAQ89066.1 | |||
| BC031265.1 | AAH31265.1 | |||
| BF338835.1 |
More Information
Set Membership: |
|

Genomic Map