Assay Details
Target Gene Details
Entrez Gene ID: | 57863 |
Gene Name: | cell adhesion molecule 3 |
Gene Aliases: |
BIgR, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3 |
Location: |
Chr.1:159171587-159203142 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CADM3 | NM_001127173.1 | 9 | 2263 | NP_001120645.1 |
| NM_021189.3 | 10 | 2402 | NP_067012.1 | |
| AF062733.2 | 10 | 2359 | AAD17540.2 | |
| AF363367.1 | 9 | 2261 | AAM60749.1 | |
| AK001560.1 | 7 | 1180 | BAA91756.1 |
Target Gene Details
Entrez Gene ID: | 100131825 |
Gene Name: | CADM3 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:159195985-159202422 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CADM3-AS1 | NR_037870.1 | |||
| AK124069.1 | BAC85765.1 |
More Information
Set Membership: |
|

Genomic Map