Assay Details
Target Gene Details
Entrez Gene ID: | 5432 |
Gene Name: | RNA polymerase II subunit C |
Gene Aliases: |
RPB3, RPB31, hRPB33, hsRPB3 |
Location: |
Chr.16:57462639-57472009 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR2C | NM_032940.2 | 9 | 1128 | NP_116558.1 |
| AJ224143.1 | 9 | 1081 | CAA11842.1 | |
| AJ224144.1 | 9 | 1081 | CAA11843.1 | |
| BC000409.2 | 9 | 1059 | AAH00409.1 | |
| BC003159.1 | 9 | 1100 | AAH03159.1 | |
| BC028157.1 | 9 | 1084 | AAH28157.1 | |
| J05448.1 | 9 | 1083 | AAA36586.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833245 | Chr.16:57434073 - 57584619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map