Assay Details
Target Gene Details
Entrez Gene ID: | 1719 |
Gene Name: | dihydrofolate reductase |
Gene Aliases: |
DHFRP1, DYR |
Location: |
Chr.5:80626226-80654981 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHFR | NM_000791.3 | 1 | 171 | NP_000782.1 |
| NM_001290354.1 | 1 | 171 | NP_001277283.1 | |
| NM_001290357.1 | 1 | 171 | NP_001277286.1 | |
| NR_110936.1 | 1 | 171 | ||
| BC003584.1 | 2 | 134 | AAH03584.2 | |
| BC071996.1 | 2 | 134 | AAH71996.1 | |
| CB988944.1 | 1 | 200 |
Target Gene Details
Entrez Gene ID: | 4437 |
Gene Name: | mutS homolog 3 |
Gene Aliases: |
DUP, MRP1 |
Location: |
Chr.5:80654648-80876815 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSH3 | NM_002439.4 | 1 | 164 | NP_002430.3 |
| AK289856.1 | 1 | 160 | ||
| BC011817.2 | 1 | 132 | AAH11817.1 | |
| BC130434.1 | 1 | 164 | ||
| BC130436.1 | 1 | 164 | ||
| J04810.1 | 1 | 145 | AAB47281.1 | |
| U61981.1 | 1 | 100 | AAB06045.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483112 | Chr.5:80551019 - 80731514 on Build GRCh38 | Loss |
|
| esv3605581 | Chr.5:80643908 - 80656286 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map