Assay Details
Target Gene Details
Entrez Gene ID: | 10479 |
Gene Name: | solute carrier family 9 member A6 |
Gene Aliases: |
MRSA, NHE6 |
Location: |
Chr.X:135974597-136047269 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A6 | NM_001042537.1 | 1 | 247 | NP_001036002.1 |
| NM_001177651.1 | 2 | 125 | NP_001171122.1 | |
| NM_006359.2 | 1 | 247 | NP_006350.1 | |
| XM_006724726.3 | 2 | 241 | XP_006724789.1 | |
| XM_011531243.2 | 2 | 101 | XP_011529545.1 | |
| XM_017029223.1 | 2 | 258 | XP_016884712.1 | |
| XM_017029224.1 | 2 | 388 | XP_016884713.1 | |
| XM_017029225.1 | 2 | 121 | XP_016884714.1 | |
| AB074255.1 | 1 | 171 | BAB72002.1 | |
| AF030409.1 | 1 | 206 | AAC39643.1 | |
| AK289775.1 | 1 | 171 | ||
| AK300475.1 | 2 | 116 | ||
| AK314562.1 | 1 | 239 | ||
| AK316255.1 | 2 | 116 | ||
| BC035029.1 | 1 | 209 | AAH35029.1 | |
| BC049169.1 | 1 | 171 | AAH49169.1 | |
| D87743.1 | 1 | 162 | BAA13449.1 | |
| DB483081.1 | 1 | 247 | ||
| DC308430.1 | 2 | 125 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758895 | Chr.X:135468612 - 136072582 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map