Assay Details
Target Gene Details
Entrez Gene ID: | 3002 |
Gene Name: | granzyme B |
Gene Aliases: |
CCPI, CGL-1, CGL1, CSP-B, CSPB, CTLA1, CTSGL1, HLP, SECT |
Location: |
Chr.14:24630954-24634226 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GZMB | NM_004131.4 | 1 | 71 | NP_004122.2 |
| XM_011536685.2 | XP_011534987.1 | |||
| AY232654.1 | AAP70244.1 | |||
| AY232655.1 | AAP70245.1 | |||
| AY232656.1 | AAP70246.1 | |||
| AY372494.1 | 1 | 112 | AAR23323.1 | |
| BC030195.1 | AAH30195.1 | |||
| BX283601.1 | 1 | 73 | ||
| J03189.1 | AAA36603.1 | |||
| J04071.1 | AAA52118.1 | |||
| M17016.1 | AAA36627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510631 | Chr.14:24575977 - 24703105 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map