Assay Details
Target Gene Details
Entrez Gene ID: | 3552 |
Gene Name: | interleukin 1 alpha |
Gene Aliases: |
IL-1A, IL1, IL1-ALPHA, IL1F1 |
Location: |
Chr.2:112773915-112785398 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1A | NM_000575.4 | 7 | 1815 | NP_000566.3 |
| AK314850.1 | ||||
| BC013142.2 | 6 | 860 | AAH13142.1 | |
| BT007014.1 | AAP35660.1 | |||
| M15329.1 | 6 | 856 | AAA59133.1 | |
| X02531.1 | 6 | 851 | CAA26371.1 | |
| X02851.1 | 7 | 909 | CAA26604.1 | |
| X56086.1 | 7 | 877 | CAA39566.1 |
More Information
Set Membership: |
|

Genomic Map