Assay Details
Target Gene Details
Entrez Gene ID: | 26503 |
Gene Name: | solute carrier family 17 member 5 |
Gene Aliases: |
AST, ISSD, NSD, SD, SIALIN, SIASD, SLD |
Location: |
Chr.6:73593378-73654014 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A5 | NM_012434.4 | 11 | 1855 | NP_036566.1 |
| XM_005248710.3 | 11 | 1877 | XP_005248767.1 | |
| XM_005248711.2 | 11 | 2109 | XP_005248768.1 | |
| XM_011535750.2 | 10 | 1665 | XP_011534052.1 | |
| XM_017010730.1 | 10 | 2021 | XP_016866219.1 | |
| XM_017010731.1 | 10 | 1871 | XP_016866220.1 | |
| AF244577.1 | 12 | 2112 | AAF97769.1 | |
| AJ387747.1 | 11 | 1997 | CAB62540.1 | |
| AK025880.1 | 5 | 768 | ||
| AK026921.1 | 3 | 520 | ||
| AK075320.1 | 12 | 1911 | BAC11546.1 | |
| BC020961.2 | 11 | 1814 | AAH20961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576178 | Chr.6:73593218 - 73711991 on Build GRCh38 | Gain |
|
| nsv527357 | Chr.6:73543543 - 73615680 on Build GRCh38 | Gain |
|
| nsv603694 | Chr.6:73565682 - 73656084 on Build GRCh38 | Loss |
|
| dgv6015n100 | Chr.6:73590773 - 73714632 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map