Assay Details
Target Gene Details
Entrez Gene ID: | 51397 |
Gene Name: | COMM domain containing 10 |
Gene Aliases: |
PTD002 |
Location: |
Chr.5:116084592-116446462 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COMMD10 | NM_001308080.1 | NP_001295009.1 | ||
| NM_016144.3 | NP_057228.1 | |||
| XM_011543458.2 | XP_011541760.1 | |||
| XM_011543460.2 | XP_011541762.1 | |||
| XM_017009553.1 | XP_016865042.1 | |||
| XM_017009555.1 | XP_016865044.1 | |||
| XM_017009556.1 | XP_016865045.1 | |||
| AF078857.1 | AAD44489.1 | |||
| AF161423.1 | AAF28983.1 | |||
| AK002147.1 | BAA92108.1 | |||
| AY542165.1 | AAS22247.1 | |||
| BC005179.1 | AAH05179.1 | |||
| BI859871.1 | ||||
| DA001572.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv599480 | Chr.5:116128903 - 116148555 on Build GRCh38 | Loss |
|
| nsv527320 | Chr.5:116046400 - 116237846 on Build GRCh38 | Loss |
|
| esv3606383 | Chr.5:116125329 - 116135554 on Build GRCh38 | Gain |
|
| esv3606380 | Chr.5:116078844 - 116301158 on Build GRCh38 | Loss |
|
| dgv5790n100 | Chr.5:116047007 - 116190041 on Build GRCh38 | Loss |
|
| esv34146 | Chr.5:115980247 - 116394222 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map