Assay Details
Target Gene Details
Entrez Gene ID: | 65992 |
Gene Name: | DDRGK domain containing 1 |
Gene Aliases: |
C20orf116, UFBP1, dJ1187M17.3 |
Location: |
Chr.20:3190350-3204697 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDRGK1 | NM_023935.2 | NP_076424.1 | ||
| AK310809.1 | ||||
| BC000643.1 | AAH00643.1 | |||
| BC007957.1 | AAH07957.1 | |||
| BC011851.1 | AAH11851.1 | |||
| CB305409.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067207 | Chr.20:3191285 - 3404426 on Build GRCh38 | Gain |
|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv528355 | Chr.20:3190691 - 3242579 on Build GRCh38 | Loss |
|
| nsv833898 | Chr.20:3147423 - 3309891 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map