Assay Details
Target Gene Details
Entrez Gene ID: | 8851 |
Gene Name: | cyclin dependent kinase 5 regulatory subunit 1 |
Gene Aliases: |
CDK5P35, CDK5R, NCK5A, p23, p25, p35, p35nck5a |
Location: |
Chr.17:32485281-32491256 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK5R1 | NM_003885.2 | 2 | 3669 | NP_003876.1 |
| XM_011525438.2 | 2 | 3656 | XP_011523740.1 | |
| XM_017025281.1 | 2 | 4551 | XP_016880770.1 | |
| AK095754.1 | 1 | 1324 | ||
| BC026347.1 | 1 | 3518 | AAH26347.1 | |
| BC030792.1 | 2 | 3675 | AAH30792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519839 | Chr.17:32448743 - 32502646 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map