Assay Details
Target Gene Details
Entrez Gene ID: | 26092 |
Gene Name: | torsin 1A interacting protein 1 |
Gene Aliases: |
LAP1, LAP1B |
Location: |
Chr.1:179882042-179920077 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR1AIP1 | NM_001267578.1 | NP_001254507.1 | ||
| NM_015602.3 | NP_056417.2 | |||
| XM_011509403.1 | XP_011507705.1 | |||
| XM_011509404.1 | XP_011507706.1 | |||
| AK001780.1 | BAA91906.1 | |||
| AK021613.1 | BAB13855.1 | |||
| AK023204.1 | BAB14461.1 | |||
| AK291495.1 | ||||
| AK310012.1 | ||||
| BC023247.1 | AAH23247.1 | |||
| BQ435161.1 | ||||
| CN360425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv473n100 | Chr.1:179814787 - 179895575 on Build GRCh38 | Gain |
|
| nsv518936 | Chr.1:179878267 - 179889198 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map