Assay Details
Target Gene Details
Entrez Gene ID: | 23101 |
Gene Name: | MCF.2 cell line derived transforming sequence-like 2 |
Gene Aliases: |
ARHGEF22 |
Location: |
Chr.3:183176116-183428990 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCF2L2 | NM_015078.3 | 22 | 2724 | NP_055893.3 |
| XM_011512585.2 | 14 | 2161 | XP_011510887.1 | |
| XM_011512586.2 | 15 | 1575 | XP_011510888.1 | |
| XM_017005943.1 | 22 | 3445 | XP_016861432.1 | |
| AB020668.1 | 19 | 2036 | BAA74884.1 | |
| AY172737.1 | 21 | 2276 | AAO19651.1 | |
| BC029074.2 | AAH29074.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893833 | Chr.3:183130677 - 183342208 on Build GRCh38 | Gain |
|
| nsv1006872 | Chr.3:183194738 - 183548501 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map