Assay Details
Target Gene Details
Entrez Gene ID: | 9398 |
Gene Name: | CD101 molecule |
Gene Aliases: |
EWI-101, IGSF2, V7 |
Location: |
Chr.1:116990423-117037481 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD101 | NM_001256106.2 | 4 | 1076 | NP_001243035.1 |
| NM_001256109.2 | 4 | 1076 | NP_001243038.1 | |
| NM_001256111.2 | 4 | 890 | NP_001243040.1 | |
| NM_004258.5 | 4 | 1076 | NP_004249.2 | |
| XM_017002846.1 | 5 | 1029 | XP_016858335.1 | |
| XM_017002847.1 | 5 | 1029 | XP_016858336.1 | |
| AK298237.1 | 4 | 865 | ||
| AK307542.1 | 4 | 1076 | ||
| BC130327.1 | 4 | 1037 | ||
| BC136484.1 | 4 | 1037 | ||
| BC171768.1 | 4 | 1037 | ||
| Z33642.1 | 4 | 1029 | CAA83923.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831159 | Chr.1:116844705 - 117040990 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map