Assay Details
Target Gene Details
Entrez Gene ID: | 65005 |
Gene Name: | mitochondrial ribosomal protein L9 |
Gene Aliases: |
L9mt |
Location: |
Chr.1:151759643-151763916 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL9 | NM_001300733.1 | 1 | 378 | NP_001287662.1 |
| NM_031420.3 | 1 | 378 | NP_113608.1 | |
| NR_125331.1 | 1 | 378 | ||
| AB049636.1 | BAB40841.1 | |||
| AK026363.1 | ||||
| AK309841.1 | ||||
| AK315459.1 | 1 | 378 | ||
| AW452832.1 | 1 | 156 | ||
| BG437768.1 |
Target Gene Details
Entrez Gene ID: | 51686 |
Gene Name: | ornithine decarboxylase antizyme 3 |
Gene Aliases: |
AZ3, OAZ-t, TISP15 |
Location: |
Chr.1:151762969-151771330 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAZ3 | NM_001134939.1 | NP_001128411.1 | ||
| NM_001301371.1 | NP_001288300.1 | |||
| AK125876.1 | ||||
| BC073949.1 | AAH73949.1 | |||
| BI830988.1 |
More Information
Set Membership: |
|

Genomic Map