Assay Details
Target Gene Details
Entrez Gene ID: | 56912 |
Gene Name: | intraflagellar transport 46 |
Gene Aliases: |
C11orf2, C11orf60, CFAP32 |
Location: |
Chr.11:118544528-118575624 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT46 | NM_001168618.1 | 12 | 1340 | NP_001162089.1 |
| NM_020153.3 | 13 | 1493 | NP_064538.3 | |
| XM_011542905.2 | 13 | 4235 | XP_011541207.1 | |
| XM_011542906.2 | 12 | 4077 | XP_011541208.1 | |
| XM_017018017.1 | 12 | 1232 | XP_016873506.1 | |
| XM_017018018.1 | 11 | 1203 | XP_016873507.1 | |
| AJ249980.1 | 12 | 1267 | CAB96537.1 | |
| AK025480.1 | 13 | 1452 | BAB15146.1 | |
| AK289521.1 | 12 | 1191 | ||
| AL136934.1 | 12 | 1175 | CAB66868.1 | |
| BC011647.2 | 12 | 1178 | AAH11647.1 | |
| BC012802.2 | 12 | 1271 | AAH12802.1 | |
| BC022856.1 | 12 | 1301 | AAH22856.1 | |
| CR457344.1 | CAG33625.1 |
Target Gene Details
Entrez Gene ID: | 84866 |
Gene Name: | transmembrane protein 25 |
Gene Aliases: |
- |
Location: |
Chr.11:118531088-118546670 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM25 | NM_001144037.1 | NP_001137509.1 | ||
| NM_001144038.1 | NP_001137510.1 | |||
| XM_017018431.1 | XP_016873920.1 | |||
| AK027305.1 | BAB55029.1 | |||
| AK075437.1 | BAC11620.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044281 | Chr.11:118503814 - 118554704 on Build GRCh38 | Gain |
|
| esv3579929 | Chr.11:118544686 - 118568073 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112784379] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map