Assay Details
Target Gene Details
Entrez Gene ID: | 2693 |
Gene Name: | growth hormone secretagogue receptor |
Gene Aliases: |
GHDP |
Location: |
Chr.3:172443291-172448456 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GHSR | NM_004122.2 | NP_004113.1 | ||
| NM_198407.2 | NP_940799.1 | |||
| AY429112.1 | AAR07907.1 | |||
| BC069068.1 | AAH69068.1 | |||
| BC069374.1 | AAH69374.1 | |||
| BC113547.1 | ||||
| U60179.1 | AAC50653.1 | |||
| U60181.1 | AAC50654.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829794 | Chr.3:172441561 - 172607903 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map