Assay Details
Target Gene Details
Entrez Gene ID: | 55664 |
Gene Name: | cell division cycle 37 like 1 |
Gene Aliases: |
CDC37B, HARC |
Location: |
Chr.9:4679553-4708399 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC37L1 | NM_017913.3 | 7 | 1391 | NP_060383.2 |
| AK000497.1 | 7 | 1386 | BAA91206.1 | |
| AK000646.1 | 7 | 1275 | BAA91304.1 | |
| AK225510.1 | 7 | 1275 | ||
| BC014133.2 | 7 | 1211 | AAH14133.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1018499 | Chr.9:4665656 - 4706366 on Build GRCh38 | Loss |
|
| nsv1015376 | Chr.9:4686223 - 4737890 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map