Assay Details
Target Gene Details
Entrez Gene ID: | 4129 |
Gene Name: | monoamine oxidase B |
Gene Aliases: |
- |
Location: |
Chr.X:43766610-43882475 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAOB | NM_000898.4 | 15 | 1844 | NP_000889.3 |
| XM_005272608.3 | 17 | 1944 | XP_005272665.1 | |
| XM_017029523.1 | 17 | 1994 | XP_016885012.1 | |
| XM_017029524.1 | 16 | 1804 | XP_016885013.1 | |
| AK294306.1 | 16 | 1762 | ||
| AK298942.1 | ||||
| BC022494.1 | 15 | 1790 | AAH22494.1 | |
| BM726427.1 | 1 | 68 | ||
| CA975968.1 | 2 | 305 | ||
| M69177.1 | 15 | 1745 | AAA59550.1 |
More Information
Set Membership: |
|

Genomic Map