Assay Details
Target Gene Details
Entrez Gene ID: | 29940 |
Gene Name: | dermatan sulfate epimerase |
Gene Aliases: |
DS-epi1, DSEP, DSEPI, EDSMC2, SART-2, SART2 |
Location: |
Chr.6:116254152-116441261 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSE | NM_001322937.1 | NP_001309866.1 | ||
| NM_001322938.1 | NP_001309867.1 | |||
| NM_001322940.1 | NP_001309869.1 | |||
| AK293872.1 |
Target Gene Details
Entrez Gene ID: | 7259 |
Gene Name: | TSPY like 1 |
Gene Aliases: |
TSPYL |
Location: |
Chr.6:116274859-116280117 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSPYL1 | NM_003309.3 | 1 | 1559 | NP_003300.1 |
| AF042181.1 | 1 | 182 | AAC62384.1 | |
| AJ420580.1 | ||||
| AK026849.1 | 1 | 420 | ||
| AL136629.1 | 1 | 1370 | CAB66564.1 | |
| BC048969.1 | 1 | 1372 | AAH48969.1 | |
| CR533535.1 | 1 | 1272 | CAG38566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576211 | Chr.6:116095308 - 116431057 on Build GRCh38 | Gain |
|
| nsv523972 | Chr.6:115958024 - 116809541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map