Assay Details
Target Gene Details
Entrez Gene ID: | 1876 |
Gene Name: | E2F transcription factor 6 |
Gene Aliases: |
E2F-6 |
Location: |
Chr.2:11444375-11466177 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| E2F6 | NM_001278275.1 | 8 | 2472 | NP_001265204.1 |
| NM_001278276.1 | 8 | 2534 | NP_001265205.1 | |
| NM_001278277.1 | 9 | 2605 | NP_001265206.1 | |
| NM_001278278.1 | 6 | 2346 | NP_001265207.1 | |
| NM_198256.3 | 7 | 2401 | NP_937987.2 | |
| NM_212540.2 | 7 | 2325 | NP_997705.1 | |
| NR_103490.1 | 5 | 2130 |
More Information
Set Membership: |
|

Genomic Map