Assay Details
Target Gene Details
Entrez Gene ID: | 55379 |
Gene Name: | leucine rich repeat containing 59 |
Gene Aliases: |
PRO1855, p34 |
Location: |
Chr.17:50381233-50397553 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC59 | NM_018509.3 | 7 | 2543 | NP_060979.2 |
| AF119857.1 | 4 | 1901 | AAF69611.1 | |
| AK025328.1 | 7 | 2543 | ||
| BC052279.1 | 7 | 2461 | AAH52279.1 | |
| BM664133.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833475 | Chr.17:50219520 - 50388833 on Build GRCh38 | Loss |
|
| nsv833476 | Chr.17:50316997 - 50473985 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map