Assay Details
Target Gene Details
Entrez Gene ID: | 51267 |
Gene Name: | C-type lectin domain family 1 member A |
Gene Aliases: |
CLEC-1, CLEC1 |
Location: |
Chr.12:10069504-10099065 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC1A | NM_001297748.1 | 5 | 1741 | NP_001284677.1 |
| NM_001297749.1 | 4 | 1564 | NP_001284678.1 | |
| NM_001297750.1 | 7 | 1938 | NP_001284679.1 | |
| NM_001297751.1 | 6 | 1882 | NP_001284680.1 | |
| NM_016511.3 | 6 | 1840 | NP_057595.2 | |
| XM_011520687.2 | 7 | 1921 | XP_011518989.1 | |
| AF200949.1 | AAF36830.1 | |||
| AK075114.1 | BAC11410.1 | |||
| AK091989.1 | ||||
| AY358587.1 | AAQ88950.1 | |||
| BC039072.1 | AAH39072.1 | |||
| BC067746.1 | 6 | 1760 | AAH67746.1 |
More Information
Set Membership: |
|

Genomic Map